2-Fluoro-1,3-diiodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

74691-500MG 74691-1G 74691-5G 74691-25G
77334-052EA 120.51 USD
77334-052 77343-828 77370-056 77382-130
2-Fluoro-1,3-diiodo-4-methylbenzene ≥97%
2-Fluoro-1,3-diiodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅FI₂
MW: 361.922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28791799
CAS Number: 1804519-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR