(2-Fluoro-3-iodo-4-methylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

74689-500MG 74689-1G 74689-5G 74689-25G
77334-050EA 91.67 USD
77334-050 77342-440 77370-054 77382-128
(2-Fluoro-3-iodo-4-methylphenyl)trimethylsilane ≥97%
(2-Fluoro-3-iodo-4-methylphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄FISi
MW: 308.207 g/mol
Storage Temperature: Ambient
CAS Number: 2918958-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR