1-Bromo-3-(cyclopropylmethoxy)-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

74613-500MG 74613-1G 74613-5G 74613-25G
77334-044EA 281.19 USD
77334-044 77358-228 77370-048 77390-898
1-Bromo-3-(cyclopropylmethoxy)-5-iodobenzene ≥97%
1-Bromo-3-(cyclopropylmethoxy)-5-iodobenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29094073
CAS Number: 1973419-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR