1,3-Dibromo-5-(2,2,2-trifluoroethoxy)benzene

Supplier: AOBChem USA

Warning

74596-500MG 74596-1G 74596-5G 74596-25G
77334-042EA 221.45 USD
77334-042 77358-224 77370-046 77390-896
1,3-Dibromo-5-(2,2,2-trifluoroethoxy)benzene
1,3-Dibromo-5-(2,2,2-trifluoroethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₅Br₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD31625256
CAS Number: 2112842-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR