1-Bromo-2-fluoro-3-methyl-4-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

74590-500MG 74590-1G 74590-5G
77334-040EA 347.11 USD
77334-040 77358-222 77382-124
1-Bromo-2-fluoro-3-methyl-4-(methylsulfonyl)benzene ≥97%
1-Bromo-2-fluoro-3-methyl-4-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂S
MW: 267.1153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765564
CAS Number: 2901082-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR