3-Formyl-2,4-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74580-500MG 74580-1G 74580-5G
77340-168EA 807.52 USD
77340-168 77358-216 77382-118
3-Formyl-2,4-dimethoxybenzoic acid ≥97%
3-Formyl-2,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₅
MW: 210.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24481204
CAS Number: 855641-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR