4-Bromo-2-fluoro-3-formyl-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74574-500MG 74574-1G 74574-5G
77336-662EA 390.37 USD
77336-662 77358-214 77382-116
4-Bromo-2-fluoro-3-formyl-5-methoxybenzoic acid ≥97%
4-Bromo-2-fluoro-3-formyl-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₆BrFO₄
Storage Temperature: Ambient
MDL Number: MFCD34765556
CAS Number: 2901102-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR