(3,5-Dichloro-2,6-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

74571-500MG 74571-1G 74571-5G 74571-25G
77334-038EA 193.64 USD
77334-038 77358-212 77370-044 77390-894
(3,5-Dichloro-2,6-difluorophenyl)(methyl)sulfane ≥97%
(3,5-Dichloro-2,6-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₂S
MW: 229.074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765554
CAS Number: 2901098-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR