1,5-Dichloro-2,4-difluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

74569-500MG 74569-1G 74569-5G 74569-25G
77334-036EA 176.13 USD
77334-036 77346-550 77370-042 77390-892
1,5-Dichloro-2,4-difluoro-3-iodobenzene ≥97%
1,5-Dichloro-2,4-difluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆HCl₂F₂I
MW: 308.879 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765553
CAS Number: 679836-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR