(5-Chloro-2,4-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

74535-1G 74535-5G 74535-25G
77340-966EA 64.89 USD
77340-966 77370-040 77382-114
(5-Chloro-2,4-dimethoxyphenyl)(methyl)sulfane ≥97%
(5-Chloro-2,4-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD34765523
CAS Number: 2710723-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR