N,N-Bis(t-Boc) 4-bromo-2,5-difluoroaniline ≥97%

Supplier: AOBChem USA

Warning

74523-1G 74523-5G 74523-25G
77341-440EA 85.49 USD
77341-440 77370-036 77382-110
N,N-Bis(t-Boc) 4-bromo-2,5-difluoroaniline ≥97%
N,N-Bis(t-Boc) 4-bromo-2,5-difluoroaniline

Precaución: for Research Use Only

Formula: C₁₆H₂₀BrF₂NO₄
MW: 408.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765519
CAS Number: 2807466-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR