3-Chloro-2,6-bis(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

74489-500MG 74489-1G 74489-5G 74489-25G
77334-032EA 281.19 USD
77334-032 77358-208 77370-034 77390-888
3-Chloro-2,6-bis(methylthio)phenol ≥97%
3-Chloro-2,6-bis(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉ClOS₂
Storage Temperature: Ambient
MDL Number: MFCD34765512
CAS Number: 2901103-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR