4-Iodo-4'-isobutoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

74472-500MG 74472-1G 74472-5G 74472-25G
77334-028EA 164.8 USD
77334-028 77346-156 77370-030 77390-884
4-Iodo-4'-isobutoxy-1,1'-biphenyl ≥97%
4-Iodo-4'-isobutoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇IO
MW: 352.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765496
CAS Number: 2807446-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR