4'-Isobutoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

74470-500MG 74470-1G 74470-5G 74470-25G
77334-026EA 182.31 USD
77334-026 77347-034 77370-028 77390-882
4'-Isobutoxy-[1,1'-biphenyl]-4-carbaldehyde ≥95%
4'-Isobutoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₂
MW: 254.324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765494
CAS Number: 1506101-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR