4'-Bromo-3-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

74446-500MG 74446-1G 74446-5G 74446-25G
77334-024EA 167.89 USD
77334-024 77346-230 77370-022 77390-880
4'-Bromo-3-methyl-1,1'-biphenyl ≥95%
4'-Bromo-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁Br
Storage Temperature: Ambient
MDL Number: MFCD06201385
CAS Number: 56961-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR