2-Chloro-1-fluoro-5-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

74444-500MG 74444-1G 74444-5G
77338-506EA 491.31 USD
77338-506 77358-198 77382-098
2-Chloro-1-fluoro-5-iodo-3-methylbenzene ≥97%
2-Chloro-1-fluoro-5-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.4704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751655
CAS Number: 1805952-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR