(2-Fluoro-4-methyl-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

74440-500MG 74440-1G 74440-25G 74440-5G
77334-022EA 300.76 USD
77334-022 77358-196 77390-878 77382-096
(2-Fluoro-4-methyl-1,3-phenylene)bis(methylsulfane) ≥97%
(2-Fluoro-4-methyl-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₉H₁₁FS₂
MW: 202.312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765476
CAS Number: 2807464-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR