1-(5-Bromo-3-chloro-2-fluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

74430-500MG 74430-1G 74430-5G 74430-25G
77334-020EA 182.31 USD
77334-020 77347-032 77370-020 77390-876
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanol ≥97%
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34765466
CAS Number: 1519669-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR