(5-Chloro-4-fluoro-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

74415-500MG 74415-1G 74415-25G 74415-5G
77334-014EA 305.91 USD
77334-014 77358-194 77390-872 77382-092
(5-Chloro-4-fluoro-1,3-phenylene)bis(methylsulfane) ≥97%
(5-Chloro-4-fluoro-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₈ClFS₂
MW: 222.731 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765457
CAS Number: 2807439-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR