1-(3-Chloro-2-fluorophenyl)-2,2-difluoroethanol ≥95%

Supplier: AOBChem USA

Warning

74409-500MG 74409-1G 74409-5G
77337-794EA 443.93 USD
77337-794 77358-190 77382-090
1-(3-Chloro-2-fluorophenyl)-2,2-difluoroethanol ≥95%
1-(3-Chloro-2-fluorophenyl)-2,2-difluoroethanol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
MW: 210.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765456
CAS Number: 1592827-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR