(3,4-Dichloro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

74408-500MG 74408-1G 74408-5G
77336-660EA 390.37 USD
77336-660 77358-188 77382-088
(3,4-Dichloro-5-methylphenyl)(methyl)sulfane ≥97%
(3,4-Dichloro-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂S
MW: 207.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787044
CAS Number: 1803845-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR