5-Chloro-2,3-difluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

74400-500MG 74400-1G 74400-5G
77334-006EA 329.6 USD
77334-006 77358-180 77382-084
5-Chloro-2,3-difluoro-4-methylbenzaldehyde ≥97%
5-Chloro-2,3-difluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD03701033
CAS Number: 2807444-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR