2,4,6-Trifluoro-3-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

74349-500MG 74349-1G 74349-5G 74349-25G
77334-002EA 100.94 USD
77334-002 77342-964 77370-004 77382-080
2,4,6-Trifluoro-3-isopropoxybenzaldehyde ≥97%
2,4,6-Trifluoro-3-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34765405
CAS Number: 2807447-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR