2,4,6-Trifluoro-3-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74347-1G 74347-5G 74347-25G
77341-890EA 106.09 USD
77341-890 77370-002 77382-078
2,4,6-Trifluoro-3-isopropoxybenzoic acid ≥97%
2,4,6-Trifluoro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765403
CAS Number: 2807465-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR