1,4-Difluoro-2-iodo-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

74319-500MG 74319-1G 74319-5G 74319-25G
77333-998EA 183.34 USD
77333-998 77347-138 77369-998 77390-862
1,4-Difluoro-2-iodo-3-methylbenzene ≥95%
1,4-Difluoro-2-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂I
MW: 254.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922623
CAS Number: 1208077-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR