1-Fluoro-3-iodo-2,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

74318-500MG 74318-1G 74318-25G 74318-5G
77333-996EA 292.52 USD
77333-996 77358-176 77390-860 77382-074
1-Fluoro-3-iodo-2,4-dimethylbenzene ≥97%
1-Fluoro-3-iodo-2,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈FI
Storage Temperature: Ambient
MDL Number: MFCD28794430
CAS Number: 26829-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR