2-Bromo-6-cyanobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74312-500MG 74312-1G 74312-5G 74312-25G
77333-994EA 87.55 USD
77333-994 77342-310 77369-996 77382-072
2-Bromo-6-cyanobenzoic acid ≥97%
2-Bromo-6-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
MW: 226.027 g/mol
Storage Temperature: Ambient
CAS Number: 1261446-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR