5-Bromo-2,3-difluoro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

74297-500MG 74297-1G 74297-5G
77336-704EA 391.4 USD
77336-704 77358-170 77382-064
5-Bromo-2,3-difluoro-4-iodobenzaldehyde ≥97%
5-Bromo-2,3-difluoro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₂BrF₂IO
MW: 346.895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765378
CAS Number: 2807464-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR