6-Bromo-2,3-difluoro-4-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74296-500MG 74296-1G 74296-5G
77340-292EA 886.83 USD
77340-292 77358-168 77382-062
6-Bromo-2,3-difluoro-4-formylbenzoic acid ≥97%
6-Bromo-2,3-difluoro-4-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrF₂O₃
MW: 265.008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765377
CAS Number: 2807443-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR