5-Fluoro-1-iodo-2,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

74164-500MG 74164-1G 74164-5G
77339-120EA 552.08 USD
77339-120 77358-164 77382-060
5-Fluoro-1-iodo-2,3-dimethoxybenzene ≥97%
5-Fluoro-1-iodo-2,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO₂
Storage Temperature: Ambient
MDL Number: MFCD28793042
CAS Number: 1806352-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR