5-Fluoro-2,3-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74159-500MG 74159-1G 74159-5G
77338-846EA 524.27 USD
77338-846 77358-162 77382-058
5-Fluoro-2,3-dimethoxybenzoic acid ≥97%
5-Fluoro-2,3-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₄
Storage Temperature: Ambient
MDL Number: MFCD19439896
CAS Number: 91407-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR