3-(Difluoromethoxy)-5-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

74130-500MG 74130-1G 74130-5G 74130-25G
77333-990EA 281.19 USD
77333-990 77358-158 77369-990 77390-858
3-(Difluoromethoxy)-5-fluorobenzoic acid ≥95%
3-(Difluoromethoxy)-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD14698508
CAS Number: 1214386-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR