4-Chloro-3-fluoro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

74126-500MG 74126-1G 74126-5G 74126-25G
77333-988EA 101.97 USD
77333-988 77343-018 77369-988 77382-052
4-Chloro-3-fluoro-5-methoxybenzaldehyde ≥97%
4-Chloro-3-fluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28639671
CAS Number: 1548015-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR