1-Chloro-2-ethyl-3-fluoro-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

74116-500MG 74116-1G 74116-5G 74116-25G
77333-986EA 221.45 USD
77333-986 77358-156 77369-986 77390-856
1-Chloro-2-ethyl-3-fluoro-4-methoxybenzene ≥97%
1-Chloro-2-ethyl-3-fluoro-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
MW: 188.627 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765332
CAS Number: 2709658-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR