1-Fluoro-2-iodo-4-isobutoxybenzene ≥95%

Supplier: AOBChem USA

Warning

74109-500MG 74109-1G 74109-5G 74109-25G
77333-984EA 144.2 USD
77333-984 77345-112 77369-984 77390-854
1-Fluoro-2-iodo-4-isobutoxybenzene ≥95%
1-Fluoro-2-iodo-4-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO
Storage Temperature: Ambient
MDL Number: MFCD34765327
CAS Number: 2807455-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR