2-Fluoro-5-isobutoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74102-500MG 74102-1G 74102-5G 74102-25G
77333-978EA 175.1 USD
77333-978 77346-548 77369-980 77390-850
2-Fluoro-5-isobutoxybenzoic acid ≥97%
2-Fluoro-5-isobutoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34765321
CAS Number: 2150978-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR