1,3-Difluoro-2-iodo-5-isobutoxybenzene ≥95%

Supplier: AOBChem USA

Warning

74098-500MG 74098-1G 74098-5G 74098-25G
77333-976EA 146.26 USD
77333-976 77345-180 77369-978 77390-848
1,3-Difluoro-2-iodo-5-isobutoxybenzene ≥95%
1,3-Difluoro-2-iodo-5-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂IO
MW: 312.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765318
CAS Number: 2918937-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR