2,6-Difluoro-4-isobutoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

74090-500MG 74090-1G 74090-5G 74090-25G
77333-974EA 281.19 USD
77333-974 77358-152 77369-976 77390-846
2,6-Difluoro-4-isobutoxybenzoic acid ≥97%
2,6-Difluoro-4-isobutoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765311
CAS Number: 2901082-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR