Methyl 5-ethyl-2-isopropoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

74077-500MG 74077-1G 74077-5G
77337-780EA 442.9 USD
77337-780 77358-148 77382-048
Methyl 5-ethyl-2-isopropoxybenzoate ≥95%
Methyl 5-ethyl-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765307
CAS Number: 2901109-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR