(3-Chloro-2-fluoro-5-isopropoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

74059-500MG 74059-1G 74059-5G 74059-25G
77333-964EA 276.04 USD
77333-964 77358-144 77369-966 77390-838
(3-Chloro-2-fluoro-5-isopropoxyphenyl)(methyl)sulfane ≥95%
(3-Chloro-2-fluoro-5-isopropoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFOS
MW: 234.718 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765294
CAS Number: 2901082-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR