(4-Fluoro-6-methyl-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

74037-500MG 74037-1G 74037-5G
77333-960EA 333.72 USD
77333-960 77358-138 77382-040
(4-Fluoro-6-methyl-1,3-phenylene)bis(methylsulfane) ≥97%
(4-Fluoro-6-methyl-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₉H₁₁FS₂
MW: 202.312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765273
CAS Number: 2901066-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR