(4-Chloro-3-fluoro-2-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

74027-500MG 74027-1G 74027-5G 74027-25G
77333-956EA 192.61 USD
77333-956 77358-136 77369-962 77390-832
(4-Chloro-3-fluoro-2-iodophenyl)(methyl)sulfane ≥97%
(4-Chloro-3-fluoro-2-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅ClFIS
MW: 302.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765265
CAS Number: 2708196-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR