4-Chloro-2-hydroxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73976-500MG 73976-1G 73976-5G
77337-696EA 436.72 USD
77337-696 77358-130 77382-038
4-Chloro-2-hydroxy-3-methylbenzoic acid ≥97%
4-Chloro-2-hydroxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
Storage Temperature: Ambient
MDL Number: MFCD22415150
CAS Number: 1824196-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR