3-Methyl-5-(methylthio)pyridin-2(1H)-one ≥95%

Supplier: AOBChem USA

Warning

73889-500MG 73889-1G 73889-5G
77340-056EA 739.54 USD
77340-056 77358-122 77382-034
3-Methyl-5-(methylthio)pyridin-2(1H)-one ≥95%
3-Methyl-5-(methylthio)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₉NOS
MW: 155.2174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765191
CAS Number: 1824286-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR