3-Ethyl-2-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73872-500MG 73872-1G 73872-5G
77339-854EA 662.29 USD
77339-854 77358-120 77382-032
3-Ethyl-2-fluoro-5-methylbenzoic acid ≥97%
3-Ethyl-2-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34765184
CAS Number: 2295873-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR