5-Ethyl-2,4-difluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

73858-500MG 73858-1G 73858-5G
77337-338EA 410.97 USD
77337-338 77358-118 77382-030
5-Ethyl-2,4-difluorobenzoic acid ≥95%
5-Ethyl-2,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34765173
CAS Number: 1895278-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR