3-Benzyl-4-(benzyloxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

73781-500MG 73781-1G 73781-5G
77338-528EA 494.4 USD
77338-528 77358-108 77382-022
3-Benzyl-4-(benzyloxy)benzoic acid ≥97%
3-Benzyl-4-(benzyloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₃
MW: 318.366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765148
CAS Number: 2918865-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR