2,5-Dibromo-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

73720-500MG 73720-1G 73720-5G
77339-802EA 645.81 USD
77339-802 77358-096 77382-012
2,5-Dibromo-3-methylbenzaldehyde ≥97%
2,5-Dibromo-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Br₂O
MW: 277.941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765123
CAS Number: 2091785-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR