2,5-Dibromo-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73718-500MG 73718-1G 73718-5G
77335-918EA 367.71 USD
77335-918 77358-094 77382-010
2,5-Dibromo-3-methylbenzoic acid ≥97%
2,5-Dibromo-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₂
MW: 293.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18379818
CAS Number: 908597-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR