3-Chloro-4-ethoxy-2-(trimethylsilyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

73655-500MG 73655-1G 73655-5G 73655-25G
77333-936EA 134.93 USD
77333-936 77344-490 77369-946 77390-814
3-Chloro-4-ethoxy-2-(trimethylsilyl)benzaldehyde ≥97%
3-Chloro-4-ethoxy-2-(trimethylsilyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClO₂Si
MW: 256.801 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765093
CAS Number: 2901085-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR